siRNA Details
| Gene Symbol: Cpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153107.2 | NP_694747.2 | 4 | 652 | ||
| GenBank mRNA | AF356844.1 | - | 4 | 561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpz| Gene Symbol: Cpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153107.2 | NP_694747.2 | 4 | 652 | ||
| GenBank mRNA | AF356844.1 | - | 4 | 561 | ||