siRNA Details
Gene Symbol: Pkhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153179.3 | NP_694819.2 | 6, 7 | 679 | ||
GenBank mRNA | AY130764.1 | - | 6, 7 | 671 | ||
AY438374.1 | - | 6, 7 | 553 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pkhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153179.3 | NP_694819.2 | 6, 7 | 679 | ||
GenBank mRNA | AY130764.1 | - | 6, 7 | 671 | ||
AY438374.1 | - | 6, 7 | 553 |