siRNA Details
| Gene Symbol: Dusp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153459.4 | NP_703189.3 | 3 | 2094 | ||
| GenBank mRNA | AK140372.1 | - | 3 | 2095 | ||
| AK158817.1 | - | 2 | 1270 | |||
| BC010207.1 | - | 3 | 1744 | 3964527 | ||
| BC025048.1 | - | 2 | 1094 | |||

Dusp7
Genomic Map