siRNA Details
| Gene Symbol: Cpne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153507.2 | NP_705727.1 | 7, 8 | 893 | ||
| GenBank mRNA | AK160238.1 | - | 7, 8 | 883 | ||
| BC022128.1 | - | 7, 8 | 854 | 5097527 | ||
| BC023348.1 | - | 2, 3 | 191 | 5012573 | ||
| BC031801.1 | - | 7, 8 | 893 | 3985959 | ||

Cpne2
Genomic Map