siRNA Details
| Gene Symbol: Rhobtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153514.5 | NP_705734.4 | 5 | 1895 | ||
| GenBank mRNA | AF420001.1 | - | 4 | 1386 | ||
| AK037927.1 | - | 4 | 2422 | |||
| AK038196.1 | - | 5 | 1987 | |||
| AK081100.1 | - | 5 | 1824 | |||
| AK122354.1 | - | 5 | 1876 | |||
| AK171295.1 | - | 11 | 2510 | |||
| BC056954.1 | - | 5 | 1500 | 5685456 | ||

Rhobtb2
Genomic Map