siRNA Details
| Gene Symbol: Erlin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153592.2 | NP_705820.1 | 7, 8 | 630 | ||
| GenBank mRNA | AK030557.1 | - | 7, 8 | 574 | ||
| BC036333.1 | - | 7, 8 | 595 | 3590904 | ||
| BC038374.1 | - | 7, 8 | 570 | 5355446 | ||

Erlin2
Genomic Map