siRNA Details
| Gene Symbol: Zfp128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153802.4 | NP_722497.1 | 4 | 760 | ||
| GenBank mRNA | AK032046.1 | - | 4 | 740 | ||
| AK035906.1 | - | 4 | 761 | |||
| AK132244.1 | - | 3 | 1015 | |||
| BC088998.1 | - | 4 | 731 | 6417536 | ||
| BC094248.1 | - | 4 | 731 | 5718665 | ||

Zfp128
Genomic Map