siRNA Details
| Gene Symbol: Zfp128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153802.4 | NP_722497.1 | 4 | 2909 | ||
| GenBank mRNA | AK032046.1 | - | 4 | 2889 | ||
| AK035906.1 | - | 4 | 2910 | |||
| BC088998.1 | - | 4 | 2880 | 6417536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp128| Gene Symbol: Zfp128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153802.4 | NP_722497.1 | 4 | 2909 | ||
| GenBank mRNA | AK032046.1 | - | 4 | 2889 | ||
| AK035906.1 | - | 4 | 2910 | |||
| BC088998.1 | - | 4 | 2880 | 6417536 | ||