siRNA Details
| Gene Symbol: Spata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170756.2 | NP_739562.1 | 3 | 1472 | ||
| GenBank mRNA | AK051741.1 | - | 3 | 1418 | ||
| AK122364.1 | - | 1 | 2114 | |||
| AK156188.1 | - | 3 | 1473 | |||
| AK162743.1 | - | 3 | 1472 | |||
| AK170356.1 | - | 3 | 1473 | |||
| AK171240.1 | - | 3 | 1446 | |||
| AK171664.1 | - | 3 | 1450 | |||
| AK171852.1 | - | 3 | 1462 | |||
| BC034597.1 | - | 3 | 1461 | 5363513 | ||

Spata2
Genomic Map