siRNA Details
| Gene Symbol: Nox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172203.2 | NP_757340.1 | 6 | 725 | ||
| GenBank mRNA | AF539799.1 | - | 6 | 579 | ||
| AK136432.1 | - | 6 | 640 | |||
| AY174116.1 | - | 3 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nox1| Gene Symbol: Nox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172203.2 | NP_757340.1 | 6 | 725 | ||
| GenBank mRNA | AF539799.1 | - | 6 | 579 | ||
| AK136432.1 | - | 6 | 640 | |||
| AY174116.1 | - | 3 | 320 | |||