siRNA Details
| Gene Symbol: Rnf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172281.2 | NP_758485.2 | 7 | 974 | ||
| GenBank mRNA | AK089805.1 | - | 7 | 906 | ||
| AK129184.1 | - | 7 | 892 | |||
| AK154982.1 | - | 7 | 905 | |||
| AK171462.1 | - | 7 | 903 | |||
| AK178704.1 | - | 1 | 32 | |||
| BC038348.1 | - | 7 | 888 | 4187484 | ||

Rnf40
Genomic Map