siRNA Details
| Gene Symbol: Rnf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172281.2 | NP_758485.2 | 2 | 283 | ||
| GenBank mRNA | AK089805.1 | - | 2 | 215 | ||
| AK129184.1 | - | 2 | 201 | |||
| AK154982.1 | - | 2 | 214 | |||
| AK171462.1 | - | 2 | 212 | |||
| BC038348.1 | - | 2 | 197 | 4187484 | ||
| BY301442.1 | - | 2 | 286 | |||

Rnf40
Genomic Map