siRNA Details
| Gene Symbol: Foxred1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291448.1 | NP_001278377.1 | 1 | 236 | ||
| NM_001291449.1 | NP_001278378.1 | 1 | 236 | |||
| NM_172291.2 | NP_758495.1 | 1 | 236 | |||
| GenBank mRNA | AK054231.1 | - | 1 | 128 | ||
| AK144759.1 | - | 1 | 162 | |||
| AK163722.1 | - | 1 | 95 | |||
| BC017604.1 | - | 1 | 189 | |||
| BC024806.1 | - | 1 | 106 | 5361053 | ||
| BC027386.1 | - | 1 | 135 | |||
| BC028892.1 | - | 1 | 151 | |||

Foxred1
Genomic Map