siRNA Details
| Gene Symbol: GRAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291824.1 | NP_001278753.1 | 3 | 415 | ||
| NM_001291825.1 | NP_001278754.1 | 2 | 428 | |||
| NM_004810.3 | NP_004801.1 | 3 | 401 | |||
| GenBank mRNA | AF042380.1 | - | 3 | 330 | ||
| AF090456.1 | - | 3 | 292 | |||
| AF102694.1 | - | 3 | 330 | |||
| AF121002.1 | - | 2 | 435 | |||
| AF129476.1 | - | 3 | 320 | |||
| AF236119.1 | - | 3 | 361 | |||
| AF236120.1 | - | 2 | 138 | |||
| AJ011736.1 | - | 3 | 346 | |||
| AK303657.1 | - | 3 | 400 | |||
| AK312830.1 | - | 3 | 400 | |||
| AY069959.1 | - | 3 | 401 | |||
| BC025692.1 | - | 3 | 335 | 5208327 | ||
| BC026002.1 | - | 2 | 397 | 4672797 | ||
| CR456498.1 | - | 3 | 199 | |||
| CR536524.1 | - | 2 | 138 | |||
| CR541849.1 | - | 2 | 138 | |||
| DC409651.1 | - | 3 | 416 | |||
| Y18051.1 | - | 3 | 174 | |||

GRAP2
Genomic Map