siRNA Details
Gene Symbol: Ccnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172301.3 | NP_758505.2 | 5 | 635 | ||
GenBank mRNA | AK136259.1 | - | 5 | 649 | ||
AK143597.1 | - | 5 | 591 | |||
AK163259.1 | - | 5 | 648 | |||
AK166686.1 | - | 5 | 649 | |||
AK167950.1 | - | 5 | 649 | |||
AK168441.1 | - | 5 | 649 | |||
AK168502.1 | - | 5 | 649 | |||
AK168993.1 | - | 5 | 646 | |||
BC011478.1 | - | 5 | 635 | 4014448 | ||
BC080202.1 | - | 4 | 422 | 3603637 | ||
BC085238.1 | - | 5 | 631 | 6337228 | ||
FJ931532.1 | - | 4 | 368 | |||
S43105.1 | - | 5 | 673 | |||
X58708.1 | - | 5 | 639 | |||
X64713.1 | - | 5 | 626 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
