siRNA Details
| Gene Symbol: Fbxl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172748.2 | NP_766336.2 | 4 | 725 | ||
| GenBank mRNA | AK028405.1 | - | 4 | 494 | ||
| AK031431.1 | - | 3 | 466 | |||
| BC059812.1 | - | 4 | 725 | 6401846 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl19| Gene Symbol: Fbxl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172748.2 | NP_766336.2 | 4 | 725 | ||
| GenBank mRNA | AK028405.1 | - | 4 | 494 | ||
| AK031431.1 | - | 3 | 466 | |||
| BC059812.1 | - | 4 | 725 | 6401846 | ||