siRNA Details
| Gene Symbol: Csgalnact1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252623.1 | NP_001239552.1 | 5, 6 | 1647 | ||
| NM_172753.5 | NP_766341.4 | 4, 5 | 1343 | |||
| GenBank mRNA | AK028688.1 | - | 4, 5 | 1266 | ||
| AK033522.1 | - | 2, 3 | 997 | |||
| AK049770.1 | - | 5, 6 | 1941 | |||
| AK080687.1 | - | 4, 5 | 1312 | |||
| AK133567.1 | - | 2, 3 | 996 | |||
| AK144089.1 | - | 1, 2 | 904 | |||
| BC057630.1 | - | 6, 7 | 1615 | |||
| BC089162.1 | - | 4, 5 | 1340 | 30470205 | ||

Csgalnact1
Genomic Map