siRNA Details
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172758.4 | NP_766346.1 | 4 | 581 | ||
GenBank mRNA | AK052548.1 | - | 4 | 582 | ||
AK171137.1 | - | 4 | 589 | |||
BC031853.1 | - | 2 | 456 | 4009949 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001003705, XM_006255067, AY294328, BC086369.
Panther Classification:
Gene Ontology Categories:
