siRNA Details
| Gene Symbol: Mthfsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166482.1 | NP_001159954.1 | 8 | 1085 | ||
| NM_172761.3 | NP_766349.1 | 8 | 1179 | |||
| GenBank mRNA | AK083317.1 | - | 8 | 1119 | ||
| AK141278.1 | - | 8 | 1014 | |||
| BC023782.1 | - | 8 | 1156 | |||
| BC026652.1 | - | 7 | 1055 | |||
| BC052066.1 | - | 7 | 2462 | 5708660 | ||

Mthfsd
Genomic Map