siRNA Details
| Gene Symbol: Zfp809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164624.1 | NP_001158096.1 | 4 | 439 | ||
| NM_172763.3 | NP_766351.3 | 4 | 448 | |||
| GenBank mRNA | AK049344.1 | - | 4 | 400 | ||
| AK089532.1 | - | 4 | 603 | |||
| BC099418.1 | - | 4 | 380 | 30525533 | ||
| BY729060.1 | - | 4 | 449 | |||
| CK389669.1 | - | 1 | 80 | |||

Zfp809
Genomic Map