siRNA Details
| Gene Symbol: Nfrkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172766.3 | NP_766354.2 | 4 | 524 | ||
| GenBank mRNA | AK036381.1 | - | 1 | 1983 | ||
| AK041206.1 | - | 4 | 520 | |||
| BC030051.1 | - | 1 | 75 | |||
| BC033595.1 | - | 4 | 514 | 4947357 | ||
| BC059861.1 | - | 4 | 503 | |||
| BC060076.1 | - | 4 | 511 | |||

Nfrkb
Genomic Map