siRNA Details
| Gene Symbol: Gramd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172768.1 | NP_766356.1 | 15 | 2109 | ||
| GenBank mRNA | AK028277.1 | - | 16 | 2004 | ||
| AK033156.1 | - | 15 | 2109 | |||
| AK122465.1 | - | 14 | 2069 | |||
| BC030911.1 | - | 6 | 853 | |||
| BC070451.1 | - | 14 | 1943 | 30616509 | ||

Gramd1b
Genomic Map