siRNA Details
| Gene Symbol: Fam63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172772.2 | NP_766360.2 | 7 | 1503 | ||
| GenBank mRNA | AK035930.1 | - | 7 | 1504 | ||
| AK046403.1 | - | 7 | 1490 | |||
| AK077470.1 | - | 4 | 435 | |||
| AK212033.1 | - | 3 | 285 | |||
| AK214619.1 | - | 2 | 171 | |||
| AK216605.1 | - | 2 | 171 | |||
| AK220158.1 | - | 7 | 1532 | |||
| BC049357.1 | - | 7 | 1480 | |||
| BC055816.1 | - | 7 | 1231 | |||
| BC058683.1 | - | 7 | 1461 | 6834871 | ||
| BC069845.1 | - | 7 | 1216 | |||

Fam63b
Genomic Map