siRNA Details
| Gene Symbol: L3mbtl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172787.2 | NP_766375.1 | 5 | 483 | ||
| GenBank mRNA | AK031398.1 | - | 5 | 469 | ||
| AK045667.1 | - | 5 | 484 | |||
| AK049597.1 | - | 5 | 518 | |||
| AK129443.1 | - | 1 | 28 | |||
| BC082309.1 | - | 4 | 353 | 30356672 | ||
| BC085192.1 | - | 4 | 588 | 30694487 | ||

L3mbtl3
Genomic Map