siRNA Details
| Gene Symbol: Sh3rf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172788.3 | NP_766376.2 | 6 | 1719 | ||
| GenBank mRNA | AK029202.1 | - | 6 | 1622 | ||
| AK132983.1 | - | 3 | 672 | |||
| BC057304.1 | - | 6 | 975 | 6830567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3rf3| Gene Symbol: Sh3rf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172788.3 | NP_766376.2 | 6 | 1719 | ||
| GenBank mRNA | AK029202.1 | - | 6 | 1622 | ||
| AK132983.1 | - | 3 | 672 | |||
| BC057304.1 | - | 6 | 975 | 6830567 | ||