siRNA Details
| Gene Symbol: Sdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172800.2 | NP_766388.2 | 39 | 5480 | ||
| GenBank mRNA | AK129379.1 | - | 35 | 4861 | ||
| AK161326.1 | - | 7 | 846 | |||
| AY351699.1 | - | 39 | 5480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdk2| Gene Symbol: Sdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172800.2 | NP_766388.2 | 39 | 5480 | ||
| GenBank mRNA | AK129379.1 | - | 35 | 4861 | ||
| AK161326.1 | - | 7 | 846 | |||
| AY351699.1 | - | 39 | 5480 | |||