siRNA Details
| Gene Symbol: Syt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172804.2 | NP_766392.2 | 5 | 953 | ||
| GenBank mRNA | AB102950.1 | - | 4 | 1285 | ||
| AK032606.1 | - | 6 | 1732 | |||
| AK137129.1 | - | 5 | 961 | |||
| BC056932.1 | - | 4 | 1063 | 5708126 | ||

Syt16
Genomic Map