siRNA Details
Gene Symbol: Syt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172804.2 | NP_766392.2 | 7 | 2326 | ||
GenBank mRNA | AB102950.1 | - | 6 | 2658 | ||
AK032606.1 | - | 8 | 3105 | |||
AK138302.1 | - | 520 | ||||
AV348245.2 | - | 1 | 296 | |||
BC056932.1 | - | 6 | 2436 | 5708126 |
