siRNA Details
| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172815.3 | NP_766403.1 | 5 | 1421 | ||
| GenBank mRNA | AK049891.1 | - | 5 | 1172 | ||
| AK052457.1 | - | 4 | 1545 | |||
| AK087485.1 | - | 5 | 1422 | |||
| BC052844.1 | - | 5 | 772 | |||

Rspo2
Genomic Map