siRNA Details
| Gene Symbol: Dip2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159361.1 | NP_001152833.1 | 15, 16 | 1962 | ||
| NM_172819.3 | NP_766407.2 | 10, 11 | 1174 | |||
| GenBank mRNA | AK030064.1 | - | 10, 11 | 1175 | ||
| AK147562.1 | - | 15, 16 | 1898 | |||
| BC023757.1 | - | 1, 2 | 61 | |||
| BC137857.1 | - | 10, 11 | 1161 | 8860876 | ||
| BC137858.1 | - | 10, 11 | 1161 | 8860877 | ||

Dip2b
Genomic Map