siRNA Details
| Gene Symbol: Nyap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172849.3 | NP_766437.2 | 3 | 1124 | ||
| GenBank mRNA | AK034747.1 | - | 4 | 1139 | ||
| BC082310.1 | - | 3 | 1124 | 30355797 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nyap2| Gene Symbol: Nyap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172849.3 | NP_766437.2 | 3 | 1124 | ||
| GenBank mRNA | AK034747.1 | - | 4 | 1139 | ||
| BC082310.1 | - | 3 | 1124 | 30355797 | ||