siRNA Details
| Gene Symbol: THEMIS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105556.2 | NP_001099026.1 | 4 | 1255 | ||
| NM_001286113.1 | NP_001273042.1 | 5 | 868 | |||
| NM_001286115.1 | NP_001273044.1 | 4 | 667 | |||
| XM_005246041.3 | XP_005246098.1 | 3 | 856 | |||
| XM_006711050.1 | XP_006711113.1 | 5 | 814 | |||
| XM_011542445.2 | XP_011540747.1 | 3 | 1126 | |||
| GenBank mRNA | AB050854.1 | - | 3 | 472 | ||
| AF044896.1 | - | 2 | 907 | |||
| AK303090.1 | - | 5 | 830 | |||
| AK303141.1 | - | 4 | 663 | |||

THEMIS2
Genomic Map