siRNA Details
| Gene Symbol: Dzank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172859.2 | NP_766447.2 | 21 | 3453 | ||
| GenBank mRNA | AK032182.1 | - | 21 | 3454 | ||
| AK163994.1 | - | 19 | 3191 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dzank1| Gene Symbol: Dzank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172859.2 | NP_766447.2 | 21 | 3453 | ||
| GenBank mRNA | AK032182.1 | - | 21 | 3454 | ||
| AK163994.1 | - | 19 | 3191 | |||