siRNA Details
| Gene Symbol: Zfp462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172867.3 | NP_766455.2 | 8 | 6758 | ||
| GenBank mRNA | AK031378.1 | - | 3 | 269 | ||
| AK173266.1 | - | 6 | 1071 | |||
| BC099692.1 | - | 3 | 355 | |||
| DQ355518.1 | - | 7 | 6455 | |||
| X66902.1 | - | 4 | 461 | |||

Zfp462
Genomic Map