siRNA Details
| Gene Symbol: Zfp462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172867.3 | NP_766455.2 | 11 | 7448 | ||
| GenBank mRNA | AK031378.1 | - | 6 | 959 | ||
| AK173266.1 | - | 9 | 1761 | |||
| BC099692.1 | - | 6 | 1045 | |||
| DQ355518.1 | - | 10 | 7145 | |||

Zfp462
Genomic Map