siRNA Details
Gene Symbol: Gpatch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172876.2 | NP_766464.1 | 7 | 1974 | ||
GenBank mRNA | AK053051.1 | - | 7 | 1975 | ||
BC096468.1 | - | 7 | 1976 | 3665952 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpatch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172876.2 | NP_766464.1 | 7 | 1974 | ||
GenBank mRNA | AK053051.1 | - | 7 | 1975 | ||
BC096468.1 | - | 7 | 1976 | 3665952 |