siRNA Details
| Gene Symbol: 2900026A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172884.3 | NP_766472.1 | 8 | 1742 | ||
| GenBank mRNA | AK041243.1 | - | 8 | 1722 | ||
| AK154484.1 | - | 8 | 1782 | |||
| AK171617.1 | - | 8 | 1742 | |||
| AK220411.1 | - | 6 | 1188 | |||

2900026A02Rik
Genomic Map