siRNA Details
| Gene Symbol: Styk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172891.2 | NP_766479.1 | 4 | 1336 | ||
| GenBank mRNA | AK033606.1 | - | 4 | 1336 | ||
| AK035247.1 | - | 4 | 497 | |||
| AY563053.1 | - | 4 | 491 | |||
| BC150724.1 | - | 4 | 544 | 9087635 | ||

Styk1
Genomic Map