siRNA Details
| Gene Symbol: Parp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172893.3 | NP_766481.2 | 3 | 890 | ||
| GenBank mRNA | AK036886.1 | - | 3 | 893 | ||
| AK053205.1 | - | 3 | 791 | |||
| AK156623.1 | - | 3 | 759 | |||
| AK162200.1 | - | 3 | 461 | |||
| AK206233.1 | - | 2 | 332 | |||
| BC120733.1 | - | 3 | 723 | 40129656 | ||
| BC137645.1 | - | 3 | 723 | 8860664 | ||

Parp12
Genomic Map