siRNA Details
| Gene Symbol: Kirrel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172898.3 | NP_766486.1 | 11 | 2086 | ||
| GenBank mRNA | AK049284.1 | - | 11 | 2087 | ||
| AK083069.1 | - | 9 | 2636 | |||
| BC052773.1 | - | 11 | 1549 | 6314924 | ||
| BC158021.1 | - | 11 | 1594 | 9007327 | ||

Kirrel2
Genomic Map