siRNA Details
Gene Symbol: Fsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172904.2 | NP_766492.2 | 4 | 1037 | ||
GenBank mRNA | AK028142.1 | - | 4 | 1029 | ||
AK036674.1 | - | 1003 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172904.2 | NP_766492.2 | 4 | 1037 | ||
GenBank mRNA | AK028142.1 | - | 4 | 1029 | ||
AK036674.1 | - | 1003 |