siRNA Details
| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF539744.1 | - | 13, 14 | 3026 | ||
| AK046675.1 | - | 12, 13 | 3447 | |||
| AK088010.1 | - | 12, 13 | 3263 | |||
| BC145639.1 | - | 3095 | ||||
| BC145640.1 | - | 3119 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd163l1| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF539744.1 | - | 13, 14 | 3026 | ||
| AK046675.1 | - | 12, 13 | 3447 | |||
| AK088010.1 | - | 12, 13 | 3263 | |||
| BC145639.1 | - | 3095 | ||||
| BC145640.1 | - | 3119 | ||||