siRNA Details
| Gene Symbol: Hydin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172916.2 | NP_766504.3 | 6, 7 | 784 | ||
| GenBank mRNA | AK043971.1 | - | 5, 6 | 603 | ||
| AK142981.1 | - | 6, 7 | 771 | |||
| AY173049.1 | - | 6, 7 | 784 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hydin| Gene Symbol: Hydin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172916.2 | NP_766504.3 | 6, 7 | 784 | ||
| GenBank mRNA | AK043971.1 | - | 5, 6 | 603 | ||
| AK142981.1 | - | 6, 7 | 771 | |||
| AY173049.1 | - | 6, 7 | 784 | |||