siRNA Details
| Gene Symbol: Hydin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172916.2 | NP_766504.3 | 14 | 1955 | ||
| GenBank mRNA | AK142981.1 | - | 14 | 1942 | ||
| AY173049.1 | - | 14 | 1955 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hydin| Gene Symbol: Hydin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172916.2 | NP_766504.3 | 14 | 1955 | ||
| GenBank mRNA | AK142981.1 | - | 14 | 1942 | ||
| AY173049.1 | - | 14 | 1955 | |||