siRNA Details
| Gene Symbol: Dpy19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172920.4 | NP_766508.3 | 22 | 3254 | ||
| GenBank mRNA | AK031291.1 | - | 4 | 2334 | ||
| AK143845.1 | - | 23 | 3259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpy19l1| Gene Symbol: Dpy19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172920.4 | NP_766508.3 | 22 | 3254 | ||
| GenBank mRNA | AK031291.1 | - | 4 | 2334 | ||
| AK143845.1 | - | 23 | 3259 | |||