siRNA Details
Gene Symbol: AI118078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172923.3 | NP_766511.1 | 4 | 421 | ||
GenBank mRNA | AK036040.1 | - | 4 | 421 | ||
BC068128.1 | - | 5 | 577 | 6835202 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AI118078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172923.3 | NP_766511.1 | 4 | 421 | ||
GenBank mRNA | AK036040.1 | - | 4 | 421 | ||
BC068128.1 | - | 5 | 577 | 6835202 |