siRNA Details
| Gene Symbol: Scml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172938.3 | NP_766526.3 | 5 | 897 | ||
| GenBank mRNA | AF397911.1 | - | 5 | 766 | ||
| AK034171.1 | - | 5 | 736 | |||
| AK047972.1 | - | 3 | 323 | |||
| AK220309.1 | - | 3 | 281 | |||
| BC043310.1 | - | 5 | 692 | 5067233 | ||

Scml4
Genomic Map