siRNA Details
| Gene Symbol: Scml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172938.3 | NP_766526.3 | 9 | 3940 | ||
| GenBank mRNA | AK047972.1 | - | 7 | 3366 | ||
| AK141826.1 | - | 4 | 2965 | |||
| AK172160.1 | - | 5 | 3003 | |||
| AK220309.1 | - | 7 | 3324 | |||
| BC043310.1 | - | 9 | 3743 | 5067233 | ||

Scml4
Genomic Map