siRNA Details
| Gene Symbol: Ankrd13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172945.2 | NP_766533.2 | 16 | 2502 | ||
| GenBank mRNA | AK081149.1 | - | 1439 | |||
| BC022693.1 | - | 5 | 1048 | |||
| BC039786.1 | - | 5 | 1466 | |||
| BC056928.1 | - | 9 | 1988 | |||
| BC071189.1 | - | 10 | 1588 | |||
| BC098207.1 | - | 7 | 1798 | |||
| BC151158.1 | - | 15 | 2259 | 9088046 | ||
| BC172028.1 | - | 15 | 2260 | |||

Ankrd13b
Genomic Map