siRNA Details
Gene Symbol: Slc28a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172980.2 | NP_766568.1 | 5 | 1533 | ||
GenBank mRNA | AB485921.1 | - | 5 | 832 | ||
AF079853.2 | - | 4 | 408 | |||
AK080957.1 | - | 5 | 1534 | |||
AK152776.1 | - | 4 | 468 | |||
BC055376.1 | - | 4 | 427 | 30077988 |
Gene Symbol: Gm14085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085518.1 | NP_001078987.1 | 4 | 1371 |
